Learn More
Medchemexpress LLC HY-145388 5mg , AU-15330 CAS:2380274-50-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1453885MG
Encompass_Preferred
HY-145388 5mg AU-15330 CAS: 2380274-50-8 AU-15330 is a proteolysis-targeting chimera (PROTAC) degrader of the SWI/SNF ATPase subunits, SMARCA2 and SMARCA4. AU-15330 induces potent inhibition of tumour growth in xenograft models of prostate cancer and synergizes with the AR antagonist enzalutamide. AU-15330 induces disease remission in castration-resistant prostate cancer (CRPC) models without toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9190