Learn More
TARGETMOL CHEMICALS INC AU-224 1MG
SDP
Catalog No. 502879606
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AU-224 is an orally available benzamide derivative that is a potent gastrointestinal prokinetic. Purity 98.08%
Catalog No. 50-287-9606 Supplier TARGETMOL CHEMICALS INC Supplier No. T139971MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AU-224 is an orally available benzamide derivative that is a potent gastrointestinal prokinetic. Purity 98.08%