Learn More
Nanocomposix BioReady Covalent Gold Conjugation Kit for Lateral Flow - 40 nm Gold Nanospheres
SDP
Catalog No. NC1820747
Encompass_Preferred
This kit eliminates the need to pre-screen and validate critical reagents and supplies to ensure your best chance of success. Kit components include:- BioReady carboxyl 40 nm gold particles, 20 OD- Desalting column for removal of interfering reagents (NaCl, azide, tris, glycine)- EDC and Sulfo-NHS- All buffers necessary to complete the conjugation- Step-by-step conjugation procedure and optimization- Complementary technical and assay development support available upon request; contact us at techsupport@nanocomposix.comNo experience needed! Our kit is the optimal solution for conjugation experts and anyone new to lateral flow looking to improve existing tests or develop the next novel LF assays.
Catalog No. NC1820747 Supplier Nanocomposix Supplier No. AUZR4010M
This kit eliminates the need to pre-screen and validate critical reagents and supplies to ensure your best chance of success. Kit components include:- BioReady carboxyl 40 nm gold particles, 20 OD- Desalting column for removal of interfering reagents (NaCl, azide, tris, glycine)- EDC and Sulfo-NHS- All buffers necessary to complete the conjugation- Step-by-step conjugation procedure and optimization- Complementary technical and assay development support available upon request; contact us at techsupport@nanocomposix.comNo experience needed! Our kit is the optimal solution for conjugation experts and anyone new to lateral flow looking to improve existing tests or develop the next novel LF assays.