Learn More
Electron Microscopy Sciences EMS Gold Aperture with 15 µm hole, 3 mm O.D., 0.24 mm thick
SDP
Catalog No. 50980775
Encompass_Preferred
Advanced techniques of thin metal film application permits the apertures to retain a clean hole edge during prolonged beam exposure. Ultra-thin, half-micron design discourages contaminant accumulation and stabilizes astigmatic conditions to provide longer life. Conventional mechanical cleaning involving costly down-time is no longer necessary. Because the foil is ultra thin, care must be taken when handling the gold foil apertures.
Size: 15 µm
O.D.: 3 mm
Thickness: 0.24 mm
Catalog No. 50-980-775 Supplier Electron Microscopy Sciences Supplier No. 44015AU
Advanced techniques of thin metal film application permits the apertures to retain a clean hole edge during prolonged beam exposure. Ultra-thin, half-micron design discourages contaminant accumulation and stabilizes astigmatic conditions to provide longer life. Conventional mechanical cleaning involving costly down-time is no longer necessary. Because the foil is ultra thin, care must be taken when handling the gold foil apertures.
Size: 15 µm
O.D.: 3 mm
Thickness: 0.24 mm