Learn More
Electron Microscopy Sciences Au-flat an ultrastable Cryo-EM sample support with a 45nm holey Gold alloy film on 3mm gold mesh grids. Au-flat a derivative of patented C-flat improves image quality and resolution and reduces particle movement during imaging.
SDP
Catalog No. NC3364556
Encompass_Preferred
Au-Flat is an ultrastable Cryo-EM sample support with a 45nm holey Gold alloy film on 3 mm gold mesh grids. Au-Flat is a derivative of the patented C-Flat product. Au-Flat improves your image quality and resolution with the gold alloy film, which reduces particle movement during imaging.
Catalog No. NC3364556 Supplier Electron Microscopy Sciences Supplier No. AUFT31350
Au-Flat is an ultrastable Cryo-EM sample support with a 45nm holey Gold alloy film on 3 mm gold mesh grids. Au-Flat is a derivative of the patented C-Flat product. Au-Flat improves your image quality and resolution with the gold alloy film, which reduces particle movement during imaging.