Learn More
Nanocomposix NanoXact Gold Nanospheres - Bare (Citrate) - 40 nm, 0.05 mg/mL in aq. 2 mM sodium citrate, 100 mL
SDP
Catalog No. 501056796
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.40 nm spherical gold nanoparticles combine strong plasmonic optical properties with versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from immunodiagnostics, biosensors, sensing, optical effects, drug delivery, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. 50-105-6796 Supplier Nanocomposix Supplier No. AUCN40100M
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.40 nm spherical gold nanoparticles combine strong plasmonic optical properties with versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from immunodiagnostics, biosensors, sensing, optical effects, drug delivery, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.