Learn More
Electron Microscopy Sciences Au/Pd(60:40) Sputter Coater Target, 60 mm diameter, 0.1 mm thick
SDP
Catalog No. 5028856
Encompass
Targets have a standard thickness of 0.1 mm (100 micron) or use on most sputtering units and are available mounted or unmounted. Made from high purity metals. When ordering, please state make and model of the sputtering unit to ensure proper fit.
Diameter: 60 mm
Thickness: 0.1 mm
Catalog No. 50-288-56 Supplier Electron Microscopy Sciences Supplier No. 91011
Targets have a standard thickness of 0.1 mm (100 micron) or use on most sputtering units and are available mounted or unmounted. Made from high purity metals. When ordering, please state make and model of the sputtering unit to ensure proper fit.
Diameter: 60 mm
Thickness: 0.1 mm