Learn More
Description
Put the power of versatility to work for your lab with a smart and efficient clinical chemistry analyzer solution. The AU480 chemistry analyzer is the ideal primary clinical chemistry analyzer for low- to mid-volume hospitals and laboratories, or dedicated specialty chemistry or STAT analyzer for larger laboratories. With throughput of up to 400 photometric tests per hour (up to 800 with ISEs), increased onboard testing, reduced sample volume and easy operation, the AU480 delivers efficiency for laboratories around the world.
Robust testing capabilities
- Broaden your test offerings with more than 120 different Beckman Coulter system reagent applications available
- Conduct quick priority testing using the AU 480 clinical chemistry analyzer's STAT wheel
- Turn around critical samples fast with sample pre-dilution and auto-repeat of over-range results
Efficient reagent and consumable use
- Minimize hands-on time and reduce daily reagent inventory tasks with ready-to-use liquid concentrated reagents
- Experience lower total cost of ownership with low consumable requirements
Convenient operational features
- Provide the right operator support when needed using context-sensitive help screens and instructional videos
- Simplify operations with user-friendly graphical user interface design and touch-screen monitor
- Reduce tech-to-tech variation and achieve consistent result quality with AU family of chemistry analyzers' standardized processes
Specifications
Specifications
| Type | Clinical Chemistry Analyzer |
| Sample Type | Serum, Plasma, Urine, Whole Blood |
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Includes | ISE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.