Learn More
ATCC AU-565; Breast Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384176
Encompass
AU565 [AU-565] is an epithelial cell that was isolated from a pleural effusion of a 43-year-old, White, female, patient with breast adenocarcinoma. The cells are useful as models of cellular growth to study the differentiation of breast cancer cells.
Catalog No. 50-238-4176 Supplier ATCC Supplier No. CRL2351
AU565 [AU-565] is an epithelial cell that was isolated from a pleural effusion of a 43-year-old, White, female, patient with breast adenocarcinoma. The cells are useful as models of cellular growth to study the differentiation of breast cancer cells.