Learn More
Medchemexpress LLC Aucubin | 479-98-1 | 346.33 | 1 ML
SDP
Catalog No. 5000388467
Encompass_Preferred
Aucubin is an iridoid glycoside component found in E. ulmoides that can cross the blood-brain barrier (BBB). It exhibits anti-anxiety, antidepressive, and anti-neuroinflammatory effects.
  • Possesses antioxidant, anti-aging, and anti-inflammatory properties
  • Exhibits antimicrobial, anti-fibrotic, and anti-cancer activities
  • Offers hepatoprotective, neuroprotective, and osteoprotective benefits
  • Intended for research use only
Catalog No. 50-003-88467 Supplier Medchemexpress LLC Supplier No. HYN066410MM/1ML
Aucubin is an iridoid glycoside component found in E. ulmoides that can cross the blood-brain barrier (BBB). It exhibits anti-anxiety, antidepressive, and anti-neuroinflammatory effects.
  • Possesses antioxidant, anti-aging, and anti-inflammatory properties
  • Exhibits antimicrobial, anti-fibrotic, and anti-cancer activities
  • Offers hepatoprotective, neuroprotective, and osteoprotective benefits
  • Intended for research use only