Learn More
Medchemexpress LLC Aucubin | 479-98-1 | 346.33 | 5 MG
SDP
Catalog No. 5000388401
Encompass_Preferred
Aucubin is an iridoid glycoside component found in E. ulmoides. It can cross the blood-brain barrier (BBB) and exhibits various biological activities including anti-anxiety, antidepressive, and anti-neuroinflammatory effects.
  • Antioxidant properties
  • Anti-aging properties
  • Anti-inflammatory properties
  • Antimicrobial properties
  • Anti-fibrotic properties
  • Anti-cancer properties
  • Hepatoprotective effects
  • Neuroprotective effects
  • Osteoprotective effects
Catalog No. 50-003-88401 Supplier Medchemexpress LLC Supplier No. HYN06645MG
Aucubin is an iridoid glycoside component found in E. ulmoides. It can cross the blood-brain barrier (BBB) and exhibits various biological activities including anti-anxiety, antidepressive, and anti-neuroinflammatory effects.
  • Antioxidant properties
  • Anti-aging properties
  • Anti-inflammatory properties
  • Antimicrobial properties
  • Anti-fibrotic properties
  • Anti-cancer properties
  • Hepatoprotective effects
  • Neuroprotective effects
  • Osteoprotective effects