Learn More
Medchemexpress LLC Aucubin | 479-98-1 | 346.33 | 50 MG
SDP
Catalog No. 5000388538
Encompass_Preferred
Aucubin (Standard) is an analytical standard, an iridoid glucoside isolated from plants such as *Plantago asiatica* and *Eucommia ulmoides*, as well as from butterfly larva. It is intended for research and analytical applications, serving as a reference standard for assay. This product is commonly utilized in qualitative, quantitative, and methodological research experiments, including techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Exhibits antioxidant, anti-aging, anti-inflammatory, antimicrobial, anti-fibrotic, anti-cancer, hepatoprotective, neuroprotective, and osteoprotective effects.
  • Can cross the blood-brain barrier (BBB).
  • Shows anti-anxiety, antidepressive, and anti-neuroinflammatory effects.
  • Inhibits IgE-induced TNF-α and IL-6 production.
  • Increases PC12 cellular viability and inhibits H2O2-induced apoptotic cell death.
Catalog No. 50-003-88538 Supplier Medchemexpress LLC Supplier No. HYN0664R50MG
Aucubin (Standard) is an analytical standard, an iridoid glucoside isolated from plants such as *Plantago asiatica* and *Eucommia ulmoides*, as well as from butterfly larva. It is intended for research and analytical applications, serving as a reference standard for assay. This product is commonly utilized in qualitative, quantitative, and methodological research experiments, including techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Exhibits antioxidant, anti-aging, anti-inflammatory, antimicrobial, anti-fibrotic, anti-cancer, hepatoprotective, neuroprotective, and osteoprotective effects.
  • Can cross the blood-brain barrier (BBB).
  • Shows anti-anxiety, antidepressive, and anti-neuroinflammatory effects.
  • Inhibits IgE-induced TNF-α and IL-6 production.
  • Increases PC12 cellular viability and inhibits H2O2-induced apoptotic cell death.