Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aucubin | 479-98-1 | 346.33 | C15H22O9 | 10 MG
SDP

Catalog No. 5000298068
Encompass_Preferred

Aucubin is an iridoid glucoside research reagent (C15H22O9; MW 346.33) used in in vitro and in vivo studies for antioxidant, anti-inflammatory, antimicrobial, anti-fibrotic, and hepatoprotective research. Provided as a 10 mg powder with documented solubility and storage guidance for experimental preparation.

  • Iridoid glucoside compound with formula C15H22O9 and molecular weight 346.33.
  • Solubility: highly soluble in water and DMSO (reported up to 100 mg/mL).
  • In vivo formulation guidance achieves ≥2.5 mg/mL in common vehicle systems.
  • Storage: powder stable at -20 °C (up to 3 years) and 4 °C (up to 2 years); solutions require colder storage.
  • Packaged as a 10 mg research reagent vial suitable for stock preparation.
  • Intended for research and analytical applications; purity not specified in available documentation.

Catalog No. 50-002-98068 Supplier Medchemexpress LLC Supplier No. HYN066410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aucubin is an iridoid glucoside research reagent (C15H22O9; MW 346.33) used in in vitro and in vivo studies for antioxidant, anti-inflammatory, antimicrobial, anti-fibrotic, and hepatoprotective research. Provided as a 10 mg powder with documented solubility and storage guidance for experimental preparation.

  • Iridoid glucoside compound with formula C15H22O9 and molecular weight 346.33.
  • Solubility: highly soluble in water and DMSO (reported up to 100 mg/mL).
  • In vivo formulation guidance achieves ≥2.5 mg/mL in common vehicle systems.
  • Storage: powder stable at -20 °C (up to 3 years) and 4 °C (up to 2 years); solutions require colder storage.
  • Packaged as a 10 mg research reagent vial suitable for stock preparation.
  • Intended for research and analytical applications; purity not specified in available documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.