Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aucubin, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014261 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4261 25 mg
11-101-4260 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4261 Supplier MilliporeSigma Supelco Supplier No. 5556125MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aucubin is a chromogene iridoid glucoside isolated from the genus, Veronica L. and exhibits a broad-spectrum of biological activities such as hepatoprotective, antitoxic, neuroprotective, antiaging, antiosteoporosis, anti-inflammatory and cardioprotective effects.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 479-98-1
Physical Form Neat
Linear Formula C15H22O9
MDL Number MFCD00136026
Synonym Aucuboside; Rhimantin
Type Analytical Standard
Quantity 25 mg
Formula Weight 346.33
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.