Learn More
Medchemexpress LLC Aucubin (standard) | 479-98-1 | 346.33 | 5 MG
SDP
Catalog No. 5000388382
Encompass_Preferred
Aucubin (Standard) is an analytical reference standard of Aucubin, an iridoid glucoside isolated from various natural sources such as *Plantago asiatica*, *Eucommia ulmoides*, *Aucuba japonica*, and butterfly larva. This product is specifically intended for research and analytical applications, serving as a reference standard for assay purposes.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applicable in techniques like HPLC, GC, and MS.
  • Exhibits diverse biological activities, including antioxidant, anti-aging, and anti-inflammatory effects.
  • Demonstrates antimicrobial, anti-fibrotic, and anti-cancer properties.
  • Offers hepatoprotective, neuroprotective, and osteoprotective benefits.
Catalog No. 50-003-88382 Supplier Medchemexpress LLC Supplier No. HYN0664R5MG
Aucubin (Standard) is an analytical reference standard of Aucubin, an iridoid glucoside isolated from various natural sources such as *Plantago asiatica*, *Eucommia ulmoides*, *Aucuba japonica*, and butterfly larva. This product is specifically intended for research and analytical applications, serving as a reference standard for assay purposes.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applicable in techniques like HPLC, GC, and MS.
  • Exhibits diverse biological activities, including antioxidant, anti-aging, and anti-inflammatory effects.
  • Demonstrates antimicrobial, anti-fibrotic, and anti-cancer properties.
  • Offers hepatoprotective, neuroprotective, and osteoprotective benefits.