Learn More
Medchemexpress LLC HY-108570 10mg Medchemexpress, AUDA CAS:479413-70-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10857010MG
Encompass_Preferred
Medchemexpress, HY-108570 10mg AUDA CAS:479413-70-2 AUDA (compound 43) is a potent soluble epoxide hydrolase (sEH) inhibitor with IC50s of 18 and 69 nM for the mouse and human sEH, respectively. AUDA has anti-inflammatory activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9637