Learn More
Medchemexpress LLC Auda | 479413-70-2 | 98.0% | 392.58 | 1 ML
SDP
Catalog No. 5000344250
Encompass_Preferred
AUDA (compound 43) is a potent soluble epoxide hydrolase (sEH) inhibitor. It exhibits anti-inflammatory activity, making it suitable for various research applications.
  • Potent soluble epoxide hydrolase (sEH) inhibitor
  • Exhibits anti-inflammatory activity
  • Suitable for research applications
Catalog No. 50-003-44250 Supplier Medchemexpress LLC Supplier No. HY108570100MG
AUDA (compound 43) is a potent soluble epoxide hydrolase (sEH) inhibitor. It exhibits anti-inflammatory activity, making it suitable for various research applications.
  • Potent soluble epoxide hydrolase (sEH) inhibitor
  • Exhibits anti-inflammatory activity
  • Suitable for research applications