Learn More
Medchemexpress LLC AUDA | 479413-70-2 | 98.0% | 392.58 | 100 MG
SDP
Catalog No. 5000344302
Encompass_Preferred
AUDA is a potent soluble epoxide hydrolase (sEH) inhibitor that exhibits anti-inflammatory activity. It is effective against both mouse and human sEH, with IC50 values of 18 nM and 69 nM respectively.
  • Potent soluble epoxide hydrolase (sEH) inhibitor
  • Exhibits anti-inflammatory activity
  • Suppresses the proliferation of rat VSMCs
  • Upregulates COX-2 expression
  • Augments the migratory ability of HCAECs
  • Increases the adhesion ability of HCAECs
  • Reduces TNF-α, MMP-9, and IL-1β expression levels
Catalog No. 50-003-44302 Supplier Medchemexpress LLC Supplier No. HY10857050MG
AUDA is a potent soluble epoxide hydrolase (sEH) inhibitor that exhibits anti-inflammatory activity. It is effective against both mouse and human sEH, with IC50 values of 18 nM and 69 nM respectively.
  • Potent soluble epoxide hydrolase (sEH) inhibitor
  • Exhibits anti-inflammatory activity
  • Suppresses the proliferation of rat VSMCs
  • Upregulates COX-2 expression
  • Augments the migratory ability of HCAECs
  • Increases the adhesion ability of HCAECs
  • Reduces TNF-α, MMP-9, and IL-1β expression levels