Learn More
Medchemexpress LLC Auda | 479413-70-2 | 98.0% | 392.58 | 250 MG
SDP
Catalog No. 5000347678
Encompass_Preferred
AUDA (compound 43) is a potent soluble epoxide hydrolase (sEH) inhibitor with IC50s of 18 nM for mouse sEH and 69 nM for human sEH. It also exhibits anti-inflammatory activity.
  • Potent soluble epoxide hydrolase (sEH) inhibitor.
  • Ic50: 18 nM (mouse sEH) and 69 nM (human sEH).
  • Possesses anti-inflammatory activity.
  • Suppresses proliferation of rat VSMCs exposed to PDGF.
  • Upregulates COX-2 expression.
  • Augments migratory ability and increases adhesion ability of HCAECs.
  • Reduces TNF-α, MMP-9, and IL-1β expression levels in vivo.
Catalog No. 50-003-47678 Supplier Medchemexpress LLC Supplier No. 250MGHY108570
AUDA (compound 43) is a potent soluble epoxide hydrolase (sEH) inhibitor with IC50s of 18 nM for mouse sEH and 69 nM for human sEH. It also exhibits anti-inflammatory activity.
  • Potent soluble epoxide hydrolase (sEH) inhibitor.
  • Ic50: 18 nM (mouse sEH) and 69 nM (human sEH).
  • Possesses anti-inflammatory activity.
  • Suppresses proliferation of rat VSMCs exposed to PDGF.
  • Upregulates COX-2 expression.
  • Augments migratory ability and increases adhesion ability of HCAECs.
  • Reduces TNF-α, MMP-9, and IL-1β expression levels in vivo.