Learn More
Enzo Life Sciences AUDA (50mg). CAS: 479413-70-2
SDP
Supplier:  Enzo Life Sciences BMLEI3920050
Encompass_Preferred
Novel inhibitor of soluble epoxide hydrolase (IC50=18nM and 69nM for the mouse and human enzymes respectively). Produces a hypotensive effect in angiotensin-infused rats and prevents and reverses cardiac hypertrophy in mouse models of cardiac hypertrophy. Alternative name: 12-(3-Adamantan-1-yl-ureido)-dodecanoic acid. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0769