Learn More
Audit™ MicroControls™ Control FD Urine Chemistry
SDP
Catalog No. 22157911
Change view
Click to view available options
Type:
Control FD Urine Chemistry, Level 1
Control FD Urine Chemistry, Level 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-157-911 Control FD Urine Chemistry, Level 2
22-157-910 Control FD Urine Chemistry, Level 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-911 Supplier Audit™ MicroControls™ Supplier No. K066210
Reference control consisting of buffered solutions
  • Analytes within this kit compose Set 1 and Set 2
  • One level
  • Freeze Dried
  • 10 x 3mL
Analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Osmolality, Phosphorus, Potassium, Sodium, Urea Nitrogen and Uric Acid
Specifications
Type Control FD Urine Chemistry, Level 2