Learn More
Audit™ MicroControls™ Linearity LQ Vitamin D
SDP
Catalog No. 22157904
Change view
Click to view available options
Quantity:
5 x 2 mL
Catalog No. Quantity
22-157-904 5 x 2 mL
1 options
Catalog No. 22-157-904 Supplier Audit™ MicroControls™ Supplier No. K729M5
Assayed quality control material consisting of five levels of human serum
  • Assay levels demonstrate linear relationship to each other for their respective analytes, reagents and instruments
  • Analytes: Vitamin D
Specifications
Type Linearity LQ Vitamin D
Quantity 5 x 2 mL