Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Audit™ MicroControls™ MicroCV™ BNP Linearity Set (Low)
SDP

Catalog No. 22157107
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-107 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-157-107 Supplier Audit™ MicroControls™ Supplier No. K708M5

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Assayed calibration verification set with human plasma based solutions

  • Simulate human plasma samples to verify and validate, Analytical Measurement Range (AMR) for testing methods such as: BNP-32 and NT-proBNP
  • Five distinct levels that demonstrate linearity between each other for their respective analytes, reagents and instruments
  • Used for all BNP-32 and NT-proBNP testing methods
  • With two years of shelf-life at 2 to 8°C
  • Quantity: 5 x 1mL

Compliance

CLIA-88 Regulations

Specifications

Certifications/Compliance CLIA-88
Content And Storage 2°C to 8°C
Detectable Analytes Brain Natriuretic Peptide-32 (BNP-32), N-Terminal Brain Natriuretic Peptide (NT-proBNP)
Type Linearity FD BNP
Intended Use Moderately complex BNP-32 and NT-proBNP testing methods
Level 5
Quantity 5 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.