Learn More
Audit™ MicroControls™ MicroCV™ ISE Linearity Set

Description
- Five distinct levels that demonstrate linearity between each other for their respective analytes, reagents and instruments
- Liquid matrix for instant use
- With two years of shelf-life at 2 to 8°C
- Quantity: 5 x 5mL
Compliance
CLIA-88 Regulations
Specifications
Specifications
| Certifications/Compliance | CLIA-88 |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Chloride, Potassium, Sodium |
| Type | Linearity LQ ISE Set |
| Level | 5 |
| Quantity | 5 x 5 mL |
| Stability | 20 Days |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.