Learn More
Audit™ MicroControls™ MicroCV™ Microalbumin/Microprotein Linearity Set

Description
- Simulate human serum samples to verify and validate, analytical measurement range (AMR) for moderately complex microalbumin and microprotein testing methods
- Five distinct levels that demonstrate linearity between each other for their respective analytes, reagents and instruments
- Liquid matrix for instant use
- With two years of shelf-life at 2 to 8°C
- Quantity: 5 x 2mL
Compliance
CLIA-88 Regulations
Specifications
Specifications
| Certifications/Compliance | CLIA-88 |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Microalbumin, Microprotein |
| Type | Linearity LQ Microalbumin or Microprotein Set |
| Level | 5 |
| Quantity | 5 x 2 mL |
| Stability | 7 Days |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.