Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Audit™ MicroControls™ MicroCV™ Procalcitonin Sets

Description
- Simulate human serum samples to verify and validate, Analytical Measurement Range (AMR) for moderately complex procalcitonin testing methods
- Five distinct levels that demonstrate linearity between each other for their respective analytes, reagents and instruments
- For procalcitonin method validation
- With two years of shelf-life at 2 to 8°C
- Available in quantity: 5 x 1mL (linearity set) or 25 x 1mL (Method Validation Set)
Compliance
CLIA-88 Regulations
Specifications
Specifications
| Certifications/Compliance | CLIA-88 |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Procalcitonin |
| Type | Linearity Set |
| For Use With (Application) | Procalcitonin method validation |
| Quantity | 5 x 1 mL |
| Form | Freeze-dried |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?