Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Audit™ MicroControls™ MicroCV™ Procalcitonin Sets
SDP

Catalog No. p-4121015
Change view
Click to view available options
Quantity:
5 x 1 mL
25 x 1 mL
Type:
Linearity Set
Method Validation Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-157-129 5 x 1 mL Linearity Set
22-157-130 25 x 1 mL Method Validation Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-157-129 Supplier Audit™ MicroControls™ Supplier No. K841M5
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Assayed calibration verification set with bovine based serum albumin solutions

  • Simulate human serum samples to verify and validate, Analytical Measurement Range (AMR) for moderately complex procalcitonin testing methods
  • Five distinct levels that demonstrate linearity between each other for their respective analytes, reagents and instruments
  • For procalcitonin method validation
  • With two years of shelf-life at 2 to 8°C
  • Available in quantity: 5 x 1mL (linearity set) or 25 x 1mL (Method Validation Set)

Compliance

CLIA-88 Regulations

Specifications

Certifications/Compliance CLIA-88
Content And Storage 2°C to 8°C
Detectable Analytes Procalcitonin
Type Linearity Set
For Use With (Application) Procalcitonin method validation
Quantity 5 x 1 mL
Form Freeze-dried
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.