Learn More
AUDIT™ MicroControls™ Linearity LQ RF/CRP

Description
- Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
- FREE and easy to use Auditor™ QC 'instant reports' data reduction software or AUDIT™ Faxback.
- AUDIT™ Ship provides unrivaled, personalized standing order autoship service.
Compliance
CLIA-88 Regulations
Specifications
Specifications
| Certifications/Compliance | CLIA-88 |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | C-Reactive Protein (CRP), Rheumatoid Factor |
| For Use With (Equipment) | Clinical Analyzers |
| Type | Verification Panel |
| Disposable | Single-use |
| For Use With (Application) | Intended to simulate human patient samples for purpose of determining linearity, calibration verification, and verification of reportable range for included analytes |
| Level | A to E |
| Packaging | Kit |
| Quantity | 5 x 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.