Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Audit™ MicroControls™ MicroCV™ Siemens Centaur™ Immunoassay (Thyroid) Linearity Set
SDP

Catalog No. 22157125
Change view
Click to view available options
Quantity:
10 x 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-125 10 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-157-125 Supplier Audit™ MicroControls™ Supplier No. K807M5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Siemens Centaur™ immunoassay systems (Thyroid)

  • Calibration verification set with human and bovine serum albumin solutions
  • Simulate human serum samples to verify and validate, Analytical Measurement Range (AMR) for testing methods such as: Free T3, Free T4, Total T3, Total T4 and TSH
  • Five distinct levels that demonstrate linearity between each other for their respective analytes, reagents and instruments
  • With two years of shelf-life at 2 to 8°C
  • Quantity:10 x 3mL

Compliance

CLIA-88 Regulations

Specifications

Certifications/Compliance CLIA-88
Content And Storage 2°C to 8°C
Detectable Analytes T3 (Free), Triiodothyroxine (T3 Total), Thyroxine, T4 (Free), Thyroxine (T4 Total), Thyroid Stimulating Hormone (TSH)
Type Linearity FD Thyroid Siemens Centaur™ Set
Level 5
Quantity 10 x 3 mL
Stability 7 Days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.