Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FD Tumor Markers Siemens Atellica/Centaur
SDP

Catalog No. 22157124
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-124 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-157-124 Supplier AUDIT™ MicroControls™ Supplier No. K805M5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Linearity FD Tumor Markers Siemens Atellica/Centaur is assayed quality control material consisting of five levels.

  • Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
  • FREE and easy to use Auditor™ QC 'instant reports' data reduction software or AUDIT™ Faxback.
  • AUDIT™ Ship provides unrivaled, personalized standing order autoship service.

Compliance

CLIA-88 Regulations

Specifications

Certifications/Compliance CLIA-88
Content And Storage 2°C to 8°C
Detectable Analytes alpha-Fetoprotein (AFP), CA-125, CA 15-3, Carcinoembryonic Antigen (CEA), PSA (Total)
For Use With (Equipment) Siemens Atellica/Centaur Analyzers
Type Verification Panel
Level A to E
Quantity 10 x 1 mL
Stability 7 days when stored at 2°C to 8°C
Form Freeze-dried
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.