Learn More
AUDIT™ MicroControls™ Linearity FD Tumor Markers

Description
- Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
- FREE and easy to use Auditor™ QC 'instant reports' data reduction software or AUDIT™ Faxback.
- AUDIT™ Ship provides unrivaled, personalized standing order autoship service.
Compliance
CLIA-88 Regulations
Specifications
Specifications
| Certifications/Compliance | CLIA-88 |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | alpha-Fetoprotein (AFP), CA-125, CA 15-3, Carcinoembryonic Antigen (CEA), PSA (Free), PSA (Total) |
| For Use With (Equipment) | Clinical Immunoassay Analyzers |
| Type | Verification Panel |
| Intended Use | Simulate Human Serum samples to verify and validate, AMR for testing methods |
| Level | A to E |
| Quantity | 10 x 1 mL |
| Stability | 7 days when stored at 2°C to 8°C |
| Form | Freeze-dried |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.