Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Audit™ MicroControls™ MicroFD™ Immunoassay Control
SDP

Catalog No. 22157139
Change view
Click to view available options
Quantity:
6 x 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-139 6 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-157-139 Supplier Audit™ MicroControls™ Supplier No. K063M6
This item is not returnable. View return policy

Tri-level, human and bovine serum albumin based control

  • Simulate human patient samples for monitoring precision of laboratory testing procedures for: digoxin, estradiol, ferritin, folate, FSH, HCG, Prolactin, etc.
  • Size: 6 x 3mL

Specifications

Color White
Content And Storage 2°C to 8°C
Detectable Analytes Cortisol, Digoxin, Estradiol, Ferritin, Folate (Vitamin B9), Follicle Stimulating Hormone (FSH), hCG, Luteinizing Hormone (LH), Progesterone, Prolactin, Total PSA, T3 (Free), Triiodothyroxine (T3 Total), Testosterone, Thyroxine, T4 (Free), Thyroxine (T4 Total), Thyroid Stimulating Hormone (TSH), Vitamin B12
Type Immunoassay Control
Intended Use Quantitate PSA, TSH, Free T4, T4, T3, hCG, LH, FSH, Progesterone, Prolactin, Estradiol, Digoxin, Ferritin, Testosterone, Vitamin B12, Folate
Level 3
Lyophilized Yes
Matrix Human Serum
pH 6.9 to 7.3
Quantity 6 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.