Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Audit™ MicroControls™ MicroLQ™ High Sensitivity C-Reactive Protein Control
SDP

Catalog No. 22157132
Change view
Click to view available options
Quantity:
6 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-132 6 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-157-132 Supplier Audit™ MicroControls™ Supplier No. K021M6

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bi-level serum-based control

  • Simulate human patient samples for monitoring precision of laboratory testing procedures for High Sensitivity C-Reactive Protein (hs-CRP)
  • Liquid, ready-to-use control
  • Size: 6 x 2mL

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes hsCRP
Type MicroLQ™ High Sensitivity C-reactive Protein Control
Quantity 6 x 2 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.