Learn More
Medchemexpress LLC HY-16617 5mg Medchemexpress, Auglurant CAS:1396337-04-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY166175MG
Encompass_Preferred
Medchemexpress, HY-16617 5mg Auglurant CAS:1396337-04-4 Auglurant (VU0424238) is a novel and selective mGlu5 antagonist with an IC 50 value of 11 nM (rat) and an IC 50 value of 14 nM (human). Auglurant (VU0424238) has an acceptable CNS penetration [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2773