Learn More
ATHENA ES Augmedium™, Powder 100
Catalog No. 5000158105
Encompass_Preferred
Augmedium™ is a medium additive which conditions cells prior to induction during a recombinant protein expression protocol. This pre-induction conditioner increases the level of chaperone proteins which can improve the fraction of product that accumulates as soluble protein. Augmedium™ is specifically intended for use with strains in which the target protein accumulates as an inclusion body or as an insoluble aggregate. Augmedium™ is supplied as a powder for preparation of 50x concentrated stock solutions. It is available in 100mL and 500mL amounts.
Catalog No. 50-001-58105 Supplier ATHENA ES Supplier No. 0123
Augmedium™ is a medium additive which conditions cells prior to induction during a recombinant protein expression protocol. This pre-induction conditioner increases the level of chaperone proteins which can improve the fraction of product that accumulates as soluble protein. Augmedium™ is specifically intended for use with strains in which the target protein accumulates as an inclusion body or as an insoluble aggregate. Augmedium™ is supplied as a powder for preparation of 50x concentrated stock solutions. It is available in 100mL and 500mL amounts.