Learn More
Alkali Scientific Augmentin [Amoxicillin, Sodium Salt/Potassium Clavulanate 5:1], 2 G
SDP
Catalog No. 5000660758
Encompass_Preferred
Amoxicillin and Clavulanate is a combination antibiotic formulation widely used in microbiological and pharmaceutical research. Amoxicillin is a beta lactam antibiotic that inhibits bacterial cell wall synthesis, while clavulanate acts as a beta lactamase inhibitor that protects amoxicillin from enzymatic degradation by resistant bacteria. This combination provides broad spectrum antibacterial activity against both Gram positive and Gram negative organisms. In laboratory applications, it is used to study bacterial resistance mechanisms, antimicrobial susceptibility, and enzyme inhibition. The formulation is also used in cell culture and microbiology research where reliable inhibition of resistant bacterial strains is required.
Catalog No. 50-006-60758 Supplier Alkali Scientific Supplier No. AUG001CP
Amoxicillin and Clavulanate is a combination antibiotic formulation widely used in microbiological and pharmaceutical research. Amoxicillin is a beta lactam antibiotic that inhibits bacterial cell wall synthesis, while clavulanate acts as a beta lactamase inhibitor that protects amoxicillin from enzymatic degradation by resistant bacteria. This combination provides broad spectrum antibacterial activity against both Gram positive and Gram negative organisms. In laboratory applications, it is used to study bacterial resistance mechanisms, antimicrobial susceptibility, and enzyme inhibition. The formulation is also used in cell culture and microbiology research where reliable inhibition of resistant bacterial strains is required.