Learn More
Alkali Scientific Augmentin [Amoxicillin, Sodium Salt/Potassium Clavulanate 5:1], 25 Grams
SDP
Catalog No. 5000661095
Encompass_Preferred
Amoxicillin and Clavulanate combination is a broad spectrum antibacterial formulation widely used in microbiological research. Amoxicillin is a beta lactam antibiotic that inhibits bacterial cell wall synthesis, while clavulanate acts as a beta lactamase inhibitor, protecting amoxicillin from enzymatic degradation by resistant bacteria. This combination enhances antibacterial effectiveness against a wider range of organisms, including those producing beta lactamase enzymes. It is commonly used in antimicrobial susceptibility testing, bacterial inhibition studies, and cell culture contamination control. Supplied in a 25 gram quantity, it is suitable for laboratory applications requiring reliable and extended spectrum antibacterial activity.
Catalog No. 50-006-61095 Supplier Alkali Scientific Supplier No. AUG003CP
Amoxicillin and Clavulanate combination is a broad spectrum antibacterial formulation widely used in microbiological research. Amoxicillin is a beta lactam antibiotic that inhibits bacterial cell wall synthesis, while clavulanate acts as a beta lactamase inhibitor, protecting amoxicillin from enzymatic degradation by resistant bacteria. This combination enhances antibacterial effectiveness against a wider range of organisms, including those producing beta lactamase enzymes. It is commonly used in antimicrobial susceptibility testing, bacterial inhibition studies, and cell culture contamination control. Supplied in a 25 gram quantity, it is suitable for laboratory applications requiring reliable and extended spectrum antibacterial activity.