Learn More
Aviva Systems Biology Corp AUH Antibody - C-terminal region (ARP40374_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40374_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AUH. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human AUH. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57417