Learn More
Aviva Systems Biology Corp Auh Antibody - N-terminal region (ARP40373_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40373_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Auh. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C77140, W91705ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-867