Learn More
ABclonal Technology AUH Rabbit mAb
SDP
Catalog No. 5000007325
Encompass_Preferred
This gene encodes bifunctional mitochondrial protein that has both RNA-binding and hydratase activities The encoded protein is a methylglutaconyl-CoA hydratase that catalyzes the hydration of 3-methylglutaconyl-CoA to 3-hydroxy-3-methyl-glutaryl-CoA a critical step in the leucine degradation pathway This protein also binds AU-rich elements (AREs) found in the 3 UTRs of rapidly decaying mRNAs including c-fos c-myc and granulocyte macrophage colony stimulating factor ARE elements are involved in directing RNA to rapid degradation and deadenylation This protein is localizes to the mitochondrial matrix and the inner mitochondrial membrane and may be involved in mitochondrial protein synthesis Mutations in this gene are the cause of 3-methylglutaconic aciduria type I Alternative splicing results in multiple transcript variants
Catalog No. 50-000-07325 Supplier ABclonal Technology Supplier No. A2412920UL
This gene encodes bifunctional mitochondrial protein that has both RNA-binding and hydratase activities The encoded protein is a methylglutaconyl-CoA hydratase that catalyzes the hydration of 3-methylglutaconyl-CoA to 3-hydroxy-3-methyl-glutaryl-CoA a critical step in the leucine degradation pathway This protein also binds AU-rich elements (AREs) found in the 3 UTRs of rapidly decaying mRNAs including c-fos c-myc and granulocyte macrophage colony stimulating factor ARE elements are involved in directing RNA to rapid degradation and deadenylation This protein is localizes to the mitochondrial matrix and the inner mitochondrial membrane and may be involved in mitochondrial protein synthesis Mutations in this gene are the cause of 3-methylglutaconic aciduria type I Alternative splicing results in multiple transcript variants