Learn More
StatLab™ Aunex Blade Remover
SDP
Catalog No. STL48CSEA
Change view
Click to view available options
Size:
# 60
Catalog No. Size
STL48CSEA # 60
1 options
Catalog No. STL48CSEA Supplier StatLab™ Supplier No. 48CSEA
Add to Cart
Add to Cart
The Aunex one-handed blade removal system allows the user to simply push and pull to remove scalpel blades without forceps or fingers.
  • Blade remover to remove larger pathology and autopsy blades, such as the #60, and #70 blades, in addition to standard blades sizes of #11 to #26
  • Holds up to 200 blades
Specifications
Product Type Dissecting Scalpel
Size # 60
Disposable Reusable
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No