Learn More
Aunex Blade Remover
SDP
Catalog No. 19200285
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
19-200-285 1
1 options
This item is not returnable. View return policy
Catalog No. 19-200-285 Supplier Cincinnati Surgical Supplier No. 48CS
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
This is the first blade remover to remove larger pathology blades, such as the #60 and #70 blades, in addition to standard blades size 10 to 26
  • Removes all standard sized surgical blades including size 60 and 70 blades
  • Can be mounted on wall or countertop
  • Holds up to 200 blades (This number will vary based on the size of the blade being removed)
Specifications
Product Type Blade Remover
For Use With (Application) Ideal for use anywhere Blades are removed from Handles, particularly in Pathology Labs; Dental, General Practice and Veterinary Offices; Mortuaries; Universities or Classroom settings
For Use With (Equipment) Surgical Blade
Description Single-handed
Quantity 1