Learn More
Selleck Chemical LLC AUNP-12
SDP
Catalog No. 5000001729
Encompass_Preferred
AUNP-12 (Aur-012 Aurigene-012 Aurigene NP-12) a new immune checkpoint modulator is an inhibitor of the PD-1 pathway
Catalog No. 50-000-01729 Supplier Selleck Chemical LLC Supplier No. S85495MG
AUNP-12 (Aur-012 Aurigene-012 Aurigene NP-12) a new immune checkpoint modulator is an inhibitor of the PD-1 pathway