Learn More
Selleck Chemical LLC AUNP-12 2mg 1353563-85-5 Aur-012, Aurigene-012, Aurigene NP-12
SDP
Supplier:  Selleck Chemical LLC S85492MG
Encompass_Preferred
AUNP-12 (Aur-012, Aurigene-012, Aurigene NP-12), a new immune checkpoint modulator, is an inhibitor of the PD-1 pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8192