Learn More
Cayman Chemical Aunt fumatuus Nautn 10mg
SDP
Catalog No. 502934477
Encompass_Preferred
A flavanone; reduces airway sensitivity, eosinophil counts and IL-4 levels in BALF, and serum IgE levels in a mouse model of ovalbumin-induced airway inflammation at 10 mg/kg; decreases immobility in the forced swim test and latency to feeding in the novelty-suppressed feeding test, indicating antidepressant-like activity in mice
Catalog No. 50-293-4477 Supplier Cayman Chemical Supplier No. 2512410 MG
A flavanone; reduces airway sensitivity, eosinophil counts and IL-4 levels in BALF, and serum IgE levels in a mouse model of ovalbumin-induced airway inflammation at 10 mg/kg; decreases immobility in the forced swim test and latency to feeding in the novelty-suppressed feeding test, indicating antidepressant-like activity in mice