Learn More
Cayman Chemical Narirutin, Purity: ≥98%, CAS: 14259-46-2, 50 mg
SDP
Catalog No. 502933497
Encompass_Preferred
A flavanone; reduces airway sensitivity, eosinophil counts and IL-4 levels in BALF, and serum IgE levels in a mouse model of ovalbumin-induced airway inflammation at 10 mg/kg; decreases immobility in the forced swim test and latency to feeding in the novelty-suppressed feeding test, indicating antidepressant-like activity in mice
Catalog No. 50-293-3497 Supplier Cayman Chemical Supplier No. 2512450 MG
A flavanone; reduces airway sensitivity, eosinophil counts and IL-4 levels in BALF, and serum IgE levels in a mouse model of ovalbumin-induced airway inflammation at 10 mg/kg; decreases immobility in the forced swim test and latency to feeding in the novelty-suppressed feeding test, indicating antidepressant-like activity in mice