Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Accessory, HEPA Filters
Catalog No. 07001180 Shop All Mystaire Products
Change view
Click to view available options
For Use With (Equipment):
Aura Elite 30 and 54 Ductless Fume Hood
Aura Elite 30 and 54 Ductless Fume Hood (Safety)
Aura Elite 42 Ductless Fume Hood
Aura Elite 42 Ductless Fume Hood (Safety)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
07-001-180 -
15-338-454 Aura Elite 42 Ductless Fume Hood
15-338-423 Aura Elite 30 and 54 Ductless Fume Hood
15-338-436 Aura Elite 42 Ductless Fume Hood (Safety)
15-338-437 Aura Elite 30 and 54 Ductless Fume Hood (Safety)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-001-180 Supplier Mystaire™ Supplier No. 657896
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful airborne particulate.
  • Provides 99.97% efficient down to 0.3μ particles