Learn More
3M™ N95 Aura™ Health Care Particulate Respirator and Surgical Mask, 1870+
Catalog No. 19072339 Shop All 3M Products
Change view
Click to view available options
Design Type:
Flat-fold
Catalog No. Design Type
19-072-339 Flat-fold
1 options
Catalog No. 19-072-339 Supplier 3M™ Supplier No. 7100235156
  • $1,113.00
    $919.65 / Case of 440
    Save $193.35
    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
 Restricted Item
Add to Cart
Add to Cart
Comfortable inner materials provide respiratory protection against certain airborne particles. Individually packaged flat-fold design allows for convenient storage.
  • Healthcare respirator meets CDC guidelines for M. tuberculosis exposure control
  • Disposable particulate respirator intended to reduce exposure to airborne particles including those generated by electrocautery, laser surgery and other powered medical instruments
  • Surgical mask designed to be fluid-resistant to splash and spatter of blood and other infectious materials
Note:
  • Before use, wearer must read and understand user instructions provided as part of the product packaging
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Flat-fold
Disposable Single-use
Filter Type N95
Material Polypropylene
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Standard
With Exhalation Valve No
Model 7100235156
Packaging Type Individually wrapped
Resistant To Fluid 160 mmHg
Size (English) 5.25 tall x 5.75 wide x 3 in. deep (approx.)
With Faceseal No
FDA Cleared Surgical Mask Yes