Learn More
3M™ Aura™ Series N95 Particulate Respirators
Catalog No. 19072338 Shop All 3M Products
Change view
Click to view available options
N95 Particulate Respirator Aura™ Model 9210+
Catalog No.
19-072-338 N95 Particulate Respirator Aura™ Model 9210+
1 options
Catalog No. 19-072-338 Supplier 3M™ Supplier No. 7100230348
Particulate respirators with or without Cool Flow™ valve offer a comfortable fit that conform to a wide range of face sizes.
  • Available with or without Cool Flow™ one-way exhalation valve for easy exhalation and cool, dry comfort in work settings that involve heat, humidity, or long periods of wear
  • Three-panel construction resists collapse and provides better visibility
  • Soft cover web on inner panel feels soft against skin
  • Standard adjustable nose clip for custom fit and secure seal
Applications:
Grinding, bagging, sanding, sweeping and other dusty operations
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0007
Certifications (OSHA) OSHA 1910.134
Design Type Flat-fold
Disposable Single-use
Filter Type N95
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve No
Filter Efficiency 95%
Latex Free Yes
Model 9210-PLUS
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Packaging Type Individually wrapped
Sterility Non-sterile
With Faceseal No
Strap Attachment Type Stapled Braided Headbands
FDA Cleared Surgical Mask No