Learn More
Medchemexpress LLC Aurachin D | 108354-13-8 | 5 MG
SDP
Catalog No. 5000418049
Encompass_Preferred
Aurachin D is an antibiotic that inhibits gram-positive bacteria and certain fungi. It also blocks NADH oxidation in beef heart submitochondrial particles. This laboratory chemical, with molecular formula C25H33NO and molecular weight 363.54, appears as a solid and is intended for research use only.
  • Inhibits gram-positive bacteria
  • Inhibits certain fungi
  • Blocks NADH oxidation in beef heart submitochondrial particles
  • Acts as an antibiotic targeting bacterial and fungal infections
  • Exhibits cytotoxicity against mouse L929 cells
Catalog No. 50-004-18049 Supplier Medchemexpress LLC Supplier No. HYN71515MG
Aurachin D is an antibiotic that inhibits gram-positive bacteria and certain fungi. It also blocks NADH oxidation in beef heart submitochondrial particles. This laboratory chemical, with molecular formula C25H33NO and molecular weight 363.54, appears as a solid and is intended for research use only.
  • Inhibits gram-positive bacteria
  • Inhibits certain fungi
  • Blocks NADH oxidation in beef heart submitochondrial particles
  • Acts as an antibiotic targeting bacterial and fungal infections
  • Exhibits cytotoxicity against mouse L929 cells